ENAH5802E8E5 Enamine

2-cyano-3-[4-(dimethylamino)phenyl]prop-2-enoic acid

  • MDL number MFCD00518737
  • Cas Number : 911424-37-8
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 208-209
Iupac Name (2Z)-2-cyano-3-[4-(dimethylamino)phenyl]-2-propenoic acid
Inchi Code 1S/C12H12N2O2/c1-14(2)11-5-3-9(4-6-11)7-10(8-13)12(15)16/h3-7H,1-2H3,(H,15,16)/b10-7-
Inchi Key CXZXLLVNQYUMAW-YFHOEESVSA-N

Description

Melting Point 208-209
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05905
Availability details for
Quantity
  • Availability
  • Add to Cart