ENAH5802E8E8 Enamine

2-hydroxy-3-(quinolin-6-yl)propanoic acid hydrochloride

  • MDL number MFCD32066393
  • Cas Number : 2361635-33-6
  • Molecular Weight : 253.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydroxy-3-(quinolin-6-yl)propanoic acid hydrochloride
Inchi Code 1S/C12H11NO3.ClH/c14-11(12(15)16)7-8-3-4-10-9(6-8)2-1-5-13-10;/h1-6,11,14H,7H2,(H,15,16);1H
Inchi Key YBFKKBHIVVIHOX-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431656

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7431656
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart