ENAH5802E8EA Enamine

3-phenyl-3-(1H-1,2,3-triazol-1-yl)propanoic acid

  • MDL number MFCD31924182
  • Cas Number : 2003011-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745187
Availability details for
Quantity
  • Availability
  • Add to Cart