ENAH5802E8EB Enamine

6-(2-methylpropyl)-1H-indole-2-carboxylic acid

  • MDL number MFCD08277169
  • Cas Number : 900136-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6502063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6502063
Availability details for
Quantity
  • Availability
  • Add to Cart