ENAH5802E8F0 Enamine

4-(4-hydroxyphenyl)-3-methyl-1,2-oxazole-5-carboxylic acid

  • Cas Number : 1781524-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425039
Availability details for
Quantity
  • Availability
  • Add to Cart