ENAH5802E8F1 Enamine

3-[5-(pyridin-4-yl)-1,3,4-oxadiazol-2-yl]propanoic acid

  • MDL number MFCD11107682
  • Cas Number : 933685-27-9
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[5-(4-pyridinyl)-1,3,4-oxadiazol-2-yl]propanoic acid
Inchi Code 1S/C10H9N3O3/c14-9(15)2-1-8-12-13-10(16-8)7-3-5-11-6-4-7/h3-6H,1-2H2,(H,14,15)
Inchi Key KUYOHZIIDVEMPX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442903
Availability details for
Quantity
  • Availability
  • Add to Cart