ENAH5802E8F2 Enamine

6-(propan-2-yloxy)-1H-indole-2-carboxylic acid

  • MDL number MFCD06653267
  • Cas Number : 881041-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6502457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6502457
Availability details for
Quantity
  • Availability
  • Add to Cart