ENAH5802E8F4 Enamine

3,3,3-trifluoro-2-hydroxy-2-phenylpropanoic acid

  • MDL number MFCD06208429
  • Cas Number : 55519-22-7
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 105-110
Iupac Name 3,3,3-trifluoro-2-hydroxy-2-phenylpropanoic acid
Inchi Code 1S/C9H7F3O3/c10-9(11,12)8(15,7(13)14)6-4-2-1-3-5-6/h1-5,15H,(H,13,14)
Inchi Key XXYXYHGJDXFJPH-UHFFFAOYSA-N

Description

Melting Point 105-110
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7438972

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7438972
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart