ENAH5802E8F6 Enamine

5-methoxy-4-oxo-4H-chromene-2-carboxylic acid

  • MDL number MFCD11877869
  • Cas Number : 5527-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763727
Availability details for
Quantity
  • Availability
  • Add to Cart