ENAH5802E8F7 Enamine

2-cyclobutaneamidopyridine-3-carboxylic acid

  • MDL number MFCD18855249
  • Cas Number : 1342989-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1018197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1018197
Availability details for
Quantity
  • Availability
  • Add to Cart