ENAH5802E8FB Enamine

2,2-difluoro-2-[(4-fluorophenyl)sulfanyl]acetic acid

  • MDL number MFCD24070432
  • Cas Number : 911360-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-44
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5018181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5018181
Availability details for
Quantity
  • Availability
  • Add to Cart