ENAH5802E8FC Enamine

3-{1-methyl-4-oxo-1H,2H,4H-pyrazolo[3,4-d]pyrimidin-6-yl}propanoic acid

  • MDL number MFCD26143772
  • Cas Number : 1822698-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29865
Availability details for
Quantity
  • Availability
  • Add to Cart