ENAH5802E900 Enamine

8,8-dimethyl-4-oxo-4H,5H,6H,7H,8H-furo[3,2-c]azepine-2-carboxylic acid

  • Cas Number : 2375262-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443992
Availability details for
Quantity
  • Availability
  • Add to Cart