ENAH5802E903 Enamine

2-[4-methyl-2-(1H-pyrazol-5-yl)-1,3-thiazol-5-yl]acetic acid

  • MDL number MFCD32069398

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435491
Availability details for
Quantity
  • Availability
  • Add to Cart