ENAH5802E904 Enamine

4-imino-2,5-dimethyl-1H,4H-thieno[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD26143690
  • Cas Number : 923737-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26114
Availability details for
Quantity
  • Availability
  • Add to Cart