ENAH5802E907 Enamine

5-ethyl-3-(oxan-4-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD19665109

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7428041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7428041
Availability details for
Quantity
  • Availability
  • Add to Cart