ENAH5802E908 Enamine

5-ethyl-3-(oxan-3-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD19665106
  • Cas Number : 1342036-49-0
  • Molecular Weight : 225.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-ethyl-3-tetrahydro-2H-pyran-3-yl-4-isoxazolecarboxylic acid
Inchi Code 1S/C11H15NO4/c1-2-8-9(11(13)14)10(12-16-8)7-4-3-5-15-6-7/h7H,2-6H2,1H3,(H,13,14)
Inchi Key PUAQTIOSBXHNRK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427834

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7427834
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart