ENAH5802E909 Enamine

4-cyano-1-methyl-3-phenyl-1H-pyrazole-5-carboxylic acid

  • Cas Number : 93764-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219-220
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7440367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440367
Availability details for
Quantity
  • Availability
  • Add to Cart