ENAH5802E90A Enamine

7,7-dioxo-5H,6H,8H-7lambda6-thiopyrano[3,4-b]pyridine-3-carboxylic acid

  • MDL number MFCD32069345
  • Cas Number : 2361876-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435063
Availability details for
Quantity
  • Availability
  • Add to Cart