ENAH5802E90C Enamine

7-(difluoromethoxy)-2,3-dihydro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD32066561
  • Cas Number : 2361645-35-2
  • Molecular Weight : 230.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-(difluoromethoxy)-2,3-dihydrobenzofuran-2-carboxylic acid
Inchi Code 1S/C10H8F2O4/c11-10(12)16-6-3-1-2-5-4-7(9(13)14)15-8(5)6/h1-3,7,10H,4H2,(H,13,14)
Inchi Key UXRHZTRAMBRPCI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6738105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738105
Availability details for
Quantity
  • Availability
  • Add to Cart