ENAH5802E90E Enamine

3-phenyl-1-(propan-2-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD21879957
  • Cas Number : 911067-99-7
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-isopropyl-3-phenyl-1H-pyrazole-5-carboxylic acid
Inchi Code 1S/C13H14N2O2/c1-9(2)15-12(13(16)17)8-11(14-15)10-6-4-3-5-7-10/h3-9H,1-2H3,(H,16,17)
Inchi Key BIZIGKJKZFVXTM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442406

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7442406
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart