ENAH5802E90F Enamine

3-phenyl-2-(thiophen-2-yl)prop-2-enoic acid

  • Cas Number : 104314-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12933
Availability details for
Quantity
  • Availability
  • Add to Cart