ENAH5802E913 Enamine

3-(6,7-dihydroxynaphthalen-2-yl)propanoic acid

  • MDL number MFCD32066038
  • Cas Number : 2361644-37-1
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(6,7-dihydroxynaphthalen-2-yl)propanoic acid
Inchi Code 1S/C13H12O4/c14-11-6-9-3-1-8(2-4-13(16)17)5-10(9)7-12(11)15/h1,3,5-7,14-15H,2,4H2,(H,16,17)
Inchi Key RCMBIAKOZZUFJS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432265

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432265
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart