ENAH5802E915 Enamine

1-[(4-methylphenyl)methyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD11848866
  • Cas Number : 645408-49-7
  • Molecular Weight : 232.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-methylbenzyl)cyclohexanecarboxylic acid
Inchi Code 1S/C15H20O2/c1-12-5-7-13(8-6-12)11-15(14(16)17)9-3-2-4-10-15/h5-8H,2-4,9-11H2,1H3,(H,16,17)
Inchi Key QTNOPCLVDCSKQY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431699

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart