ENAH5802E916 Enamine

1-[(2-methylphenyl)methyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD16316532
  • Cas Number : 1225791-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433522
Availability details for
Quantity
  • Availability
  • Add to Cart