ENAH5802E918 Enamine

6,7-dimethoxyquinoline-2-carboxylic acid

  • MDL number MFCD19288916

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7438796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438796
Availability details for
Quantity
  • Availability
  • Add to Cart