ENAH5802E919 Enamine

(2S)-3-methyl-2-[4-(methylsulfanyl)butanamido]butanoic acid

  • Cas Number : 1608817-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-996379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-996379
Availability details for
Quantity
  • Availability
  • Add to Cart