ENAH5802E91A Enamine

1-(2-fluorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD11890547
  • Cas Number : 1181245-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427989
Availability details for
Quantity
  • Availability
  • Add to Cart