ENAH5802E921 Enamine

5-(4-hydroxyphenyl)-2-methyl-1,3-thiazole-4-carboxylic acid

  • Cas Number : 1510664-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7355436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355436
Availability details for
Quantity
  • Availability
  • Add to Cart