ENAH5802E925 Enamine

(2S)-3-(4-hydroxyphenyl)-2-propanamidopropanoic acid

  • MDL number MFCD13243496
  • Cas Number : 83240-72-6
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-3-(4-hydroxyphenyl)-2-(propionylamino)propanoic acid
Inchi Code 1S/C12H15NO4/c1-2-11(15)13-10(12(16)17)7-8-3-5-9(14)6-4-8/h3-6,10,14H,2,7H2,1H3,(H,13,15)(H,16,17)/t10-/m0/s1
Inchi Key RIUFKWHJUHBQAL-JTQLQIEISA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433635

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7433635
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart