ENAH5802E928 Enamine

4-(1H-indazol-1-yl)benzoic acid

  • MDL number MFCD09030184
  • Cas Number : 838820-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7432525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432525
Availability details for
Quantity
  • Availability
  • Add to Cart