ENAH5802E92C Enamine

(2S)-3-(4-hydroxyphenyl)-2-[(methoxycarbonyl)amino]propanoic acid

  • MDL number MFCD16380126
  • Cas Number : 338386-45-1
  • Molecular Weight : 239.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 89-94
Iupac Name (2S)-3-(4-hydroxyphenyl)-2-[(methoxycarbonyl)amino]propanoic acid
Inchi Code 1S/C11H13NO5/c1-17-11(16)12-9(10(14)15)6-7-2-4-8(13)5-3-7/h2-5,9,13H,6H2,1H3,(H,12,16)(H,14,15)/t9-/m0/s1
Inchi Key FLXYBPYOUFSWKY-VIFPVBQESA-N

Description

Melting Point 89-94
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430850

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7430850
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart