ENAH5802E92D Enamine

7-phenylpyrazolo[1,5-a]pyrimidine-5-carboxylic acid

  • MDL number MFCD32639672
  • Cas Number : 1782684-07-4
  • Molecular Weight : 239.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-phenylpyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Inchi Code 1S/C13H9N3O2/c17-13(18)10-8-11(9-4-2-1-3-5-9)16-12(15-10)6-7-14-16/h1-8H,(H,17,18)
Inchi Key HHCJRQYACGHKCK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436114

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7436114
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart