ENAH5802E92E Enamine

5-(quinolin-2-yl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD32066432
  • Cas Number : 2355724-22-8
  • Molecular Weight : 240.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431785
Availability details for
Quantity
  • Availability
  • Add to Cart