ENAH5802E930 Enamine

6-chloro-4-hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline-4-carboxylic acid

  • MDL number MFCD32069411
  • Cas Number : 2375261-35-9
  • Molecular Weight : 241.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4-hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline-4-carboxylic acid

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436622
Availability details for
Quantity
  • Availability
  • Add to Cart