ENAH5802E931 Enamine

2-chloro-5-sulfamoylthiophene-3-carboxylic acid

  • MDL number MFCD01859821
  • Cas Number : 154064-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7354747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354747
Availability details for
Quantity
  • Availability
  • Add to Cart