ENAH5802E934 Enamine

2-methyl-2-(5-methyl-1-phenyl-1H-pyrazol-3-yl)propanoic acid

  • MDL number MFCD32197964
  • Cas Number : 1893564-19-6
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-2-(5-methyl-1-phenyl-1H-pyrazol-3-yl)propanoic acid
Inchi Code 1S/C14H16N2O2/c1-10-9-12(14(2,3)13(17)18)15-16(10)11-7-5-4-6-8-11/h4-9H,1-3H3,(H,17,18)
Inchi Key FBMVOPGODKVPQU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442486

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7442486
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart