ENAH5802E936 Enamine

3-(3-methylphenyl)-2-(thiophen-2-yl)prop-2-enoic acid

  • Cas Number : 852400-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12777
Availability details for
Quantity
  • Availability
  • Add to Cart