ENAH5802E938 Enamine

2-methyl-5-[(1R,2S,3S)-1,2,3,4-tetrahydroxybutyl]furan-3-carboxylic acid

  • MDL number MFCD32639854
  • Molecular Weight : 246.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-5-((1R,2S,3S)-1,2,3,4-tetrahydroxybutyl)furan-3-carboxylic acid

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7356659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356659
Availability details for
Quantity
  • Availability
  • Add to Cart