ENAH5802E93A Enamine

(2S)-3-methyl-2-[5-(methylsulfanyl)pentanamido]butanoic acid

  • MDL number MFCD32197773
  • Cas Number : 1825160-93-7
  • Molecular Weight : 247.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-(methylthio)pentanoyl)-L-valine
Inchi Code 1S/C11H21NO3S/c1-8(2)10(11(14)15)12-9(13)6-4-5-7-16-3/h8,10H,4-7H2,1-3H3,(H,12,13)(H,14,15)/t10-/m0/s1
Inchi Key FNCHLEACBNBJNI-JTQLQIEISA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440069

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7440069
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart