ENAH5802E93B Enamine

2-(3,5-dichlorophenoxy)butanoic acid

  • MDL number MFCD12067247
  • Cas Number : 39644-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-118
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-6763624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763624
Availability details for
Quantity
  • Availability
  • Add to Cart