ENAH5802E93E Enamine

4-benzyl-5,5-dimethylmorpholine-3-carboxylic acid hydrochloride

  • Cas Number : 2375259-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443876
Availability details for
Quantity
  • Availability
  • Add to Cart