ENAH5802E940 Enamine

3-(2,4-difluorophenoxy)benzoic acid

  • MDL number MFCD19551754

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7430490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430490
Availability details for
Quantity
  • Availability
  • Add to Cart