ENAH5802E946 Enamine

5-(2-chloro-4-methoxyphenyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD07377165
  • Cas Number : 1188200-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434962
Availability details for
Quantity
  • Availability
  • Add to Cart