ENAH5802E947 Enamine

4-(1,1,3,3,3-pentafluoropropyl)benzoic acid

  • MDL number MFCD32068799
  • Cas Number : 2361727-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1270467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1270467
Availability details for
Quantity
  • Availability
  • Add to Cart