ENAH5802E94D Enamine

4-[3-(propan-2-yl)phenoxy]benzoic acid

  • MDL number MFCD12735298
  • Cas Number : 1036509-09-7
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427043
Availability details for
Quantity
  • Availability
  • Add to Cart