ENAH5802E94E Enamine

2-[(2-ethylphenyl)methoxy]benzoic acid

  • MDL number MFCD32066596
  • Cas Number : 2361635-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432749
Availability details for
Quantity
  • Availability
  • Add to Cart