ENAH5802E951 Enamine

5-(2,6-dichlorophenyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD11113558
  • Cas Number : 1188024-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7352654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352654
Availability details for
Quantity
  • Availability
  • Add to Cart