ENAH5802E952 Enamine

4-(3-ethoxyphenoxy)benzoic acid

  • MDL number MFCD17527159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7427057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427057
Availability details for
Quantity
  • Availability
  • Add to Cart