ENAH5802E953 Enamine

4-(2-ethoxyphenoxy)benzoic acid

  • MDL number MFCD09706650
  • Cas Number : 938231-39-1
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-ethoxyphenoxy)benzoic acid
Inchi Code 1S/C15H14O4/c1-2-18-13-5-3-4-6-14(13)19-12-9-7-11(8-10-12)15(16)17/h3-10H,2H2,1H3,(H,16,17)
Inchi Key PJZFUDVCECUSEI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427036
Availability details for
Quantity
  • Availability
  • Add to Cart